Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MID2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109958
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109958 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109958 Supplier Invitrogen™ Supplier No. PA5109958
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GTRYIFIVKA INQAGSRNSE PTRLKTNSQP FKLDPKMTHK KLKISNDGLQ MEKDESSLKK SHTPERFSGT GCYGAAGNIF IDSGCHYWEV VMGSSTWYAI GIAYKSAPKN EWIGKNASSW VFSRCNSNFV VRHNNKEMLV DVPPHLKRLG VLLDYDNNML SFYDPANSLH LHTFDVTFIL PVCPTFTIWN KSLMILSGLP APDFIDYPER QECNCRPQES PYVSGMKTCH.

The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to microtubular structures in the cytoplasm. Alternate splicing of this gene results in two transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MID2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.45 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MID2
Gene Accession No. Q9QUS6, Q9UJV3
Gene Alias FLJ37715; FLJ41813; FXY2; Mid2; midin 2; midin-2; midline 2; midline defect 2; Midline-2; midline-2; probable E3 ubiquitin-protein ligase MID2; MRX101; probable E3 ubiquitin-protein ligase MID2; RGD1565674; RING finger protein 60; RING-type E3 ubiquitin transferase MID2; RNF60; RP6-191P20.2; Trim1; tripartite motif protein 1; tripartite motif-containing protein 1
Gene Symbols MID2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 476-705 of human MID2 (NP_438112.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11043, 23947
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.