Learn More
Invitrogen™ MID2 Polyclonal Antibody

Description
This antibody is expected to recognize both human isoforms of this protein. It is predicted to react with bovine, canine and porcine based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 4,000.
Specifications
Specifications
| Antigen | MID2 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 |
| Gene | MID2 |
| Gene Accession No. | Q9UJV3 |
| Gene Alias | FLJ37715; FLJ41813; FXY2; Mid2; midin 2; midin-2; midline 2; midline defect 2; Midline-2; midline-2; probable E3 ubiquitin-protein ligase MID2; MRX101; probable E3 ubiquitin-protein ligase MID2; RGD1565674; RING finger protein 60; RING-type E3 ubiquitin transferase MID2; RNF60; RP6-191P20.2; Trim1; tripartite motif protein 1; tripartite motif-containing protein 1 |
| Gene Symbols | MID2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.