Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
midA Antibody (OTI4A5), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272681AF700
Description
midA Monoclonal specifically detects midA in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
midA | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
NDUFAF7 | |
Full length recombinant protein of human midA (NP_653337) produced in HEK293T cells. | |
0.1 mL | |
55471.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI4A5 | |
Western Blot, Immunohistochemistry | |
chromosome 2 open reading frame 56, MidA, mitochondrial, PRO1853 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction