Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
midA Antibody (OTI4A5), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272681F
Description
midA Monoclonal specifically detects midA in Human, Mouse samples. It is validated for Immunohistochemistry.Specifications
| midA | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| NDUFAF7 | |
| Full length recombinant protein of human midA (NP_653337) produced in HEK293T cells. | |
| 0.1 mL | |
| 55471.0 | |
| Store at 4°C in the dark. |
| Immunohistochemistry | |
| OTI4A5 | |
| Immunohistochemistry | |
| chromosome 2 open reading frame 56, MidA, mitochondrial, PRO1853 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction