Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Zinc Metal, Granular, 20-Mesh, Reagent Grade, Innovating Science™

Click to view available options
Quantity:
100 g
Specifications
Specifications
Quantity | 100 g |
Packaging | HDPE |
Form | Granular |
Percent Purity | 100% |
Grade | Reagent |
Chemical Name or Material | Zinc |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction