Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Midkine Antibody (2284A), Alexa Fluor™ 700, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals FAB2581N
Description
Midkine Monoclonal antibody specifically detects Midkine in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
Midkine | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, CyTOF | |
2284A | |
Intracellular Staining by Flow Cytometry, CyTOF-ready | |
Amphiregulin-associated protein, Midgestation and kidney protein, midkine, midkine (neurite growth-promoting factor 2), MK1, MKARAP, NEGF2FLJ27379, Neurite outgrowth-promoting factor 2, Neurite outgrowth-promoting protein | |
E. coli-derived recombinant human Midkine., Lys23-Asp143, Accession # P21741 | |
0.1 mL | |
Biologically Active Proteins, Cell Cycle and Replication | |
4192 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction