Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Midkine Antibody (2284A), PE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals FAB2581P
Description
Midkine Monoclonal antibody specifically detects Midkine in Human samples. It is validated for Flow Cytometry (Intracellular Staining), CyTOFSpecifications
| Midkine | |
| Monoclonal | |
| PE | |
| PBS | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry (Intracellular Staining), CyTOF | |
| 2284A | |
| Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| Amphiregulin-associated protein, Midgestation and kidney protein, midkine, midkine (neurite growth-promoting factor 2), MK1, MKARAP, NEGF2FLJ27379, Neurite outgrowth-promoting factor 2, Neurite outgrowth-promoting protein | |
| E. coli-derived recombinant human Midkine., Lys23-Asp143, Accession # P21741 | |
| 0.1 mL | |
| Biologically Active Proteins, Cell Cycle and Replication | |
| 4192 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction