Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Midkine Antibody (OTI6C8), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272682AF405
Description
Midkine Monoclonal specifically detects Midkine in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
Midkine | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
MDK | |
Recombinant protein fragment corresponding to amino acids 21-143 of human Midkine (NP_001012334) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot | |
OTI6C8 | |
Western Blot | |
Amphiregulin-associated protein, Midgestation and kidney protein, midkine, midkine (neurite growth-promoting factor 2), MK1, MKARAP, NEGF2FLJ27379, Neurite outgrowth-promoting factor 2, Neurite outgrowth-promoting protein | |
Mouse | |
Affinity Purified | |
Biologically Active Proteins, Cell Cycle and Replication | |
4192.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction