Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Midkine Polyclonal Antibody

Catalog No. PIPA546927
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546927 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546927 Supplier Invitrogen™ Supplier No. PA546927
Add to Cart
Add to Cart

Goat Polyclonal Antibody

In direct ELISAs, approximately 10% cross-reactivity with recombinant mouse Midkine is observed, and less than 1% cross-reactivity with recombinant human Pleiotrophin is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

Midkine is a retinoic acid-responsive, heparin-binding growth factor expressed in various cell types during embryogenesis. It promotes angiogenesis, cell growth, and cell migration. Midkine is also expressed in several carcinomas, suggesting that it may play a role in tumorigenesis, perhaps through its effects on angiogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Midkine
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene Mdk
Gene Accession No. P21741
Gene Alias amphiregulin-associated protein; ARAP; FLJ27379; MDK; Mek; Midgestation and kidney protein; midkine; midkine (neurite growth-promoting factor 2); MK; MK1; NEGF2; Neurite outgrowth-promoting factor 2; Neurite outgrowth-promoting protein; Retanoic acid-responsive protein; retinoic acid inducible factor; retinoic acid-induced differentiation factor; retinoic acid-responsive protein
Gene Symbols Mdk
Host Species Goat
Immunogen E. coli-derived recombinant human Midkine Lys23-Asp143.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4192
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.