Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Midkine Polyclonal Antibody, Abbomax

Catalog No. 502203865
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-3865 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-3865 Supplier Abbomax Inc Supplier No. 600110
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive control: Full length recombinant protein Cellular location: Cytoplasm.

Midkine Polyclonal antibody specifically detects Midkine in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen Midkine
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene Mdk
Gene Accession No. P21741
Gene Alias amphiregulin-associated protein; ARAP; FLJ27379; MDK; Mek; Midgestation and kidney protein; midkine; midkine (neurite growth-promoting factor 2); MK; MK1; NEGF2; Neurite outgrowth-promoting factor 2; Neurite outgrowth-promoting protein; Retanoic acid-responsive protein; retinoic acid inducible factor; retinoic acid-induced differentiation factor; retinoic acid-responsive protein
Host Species Rabbit
Immunogen A 143 aa recombinant protein of the full length human Midkine
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4192
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.