Learn More
Invitrogen™ MIDN Polyclonal Antibody

Description
The antibody detects endogenous levels of total MIDN protein.
Specifications
Specifications
| Antigen | MIDN |
| Applications | Immunohistochemistry, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 1% BSA, 50% glycerol and 0.02% ProClin 300; pH 7.4 |
| Gene | MIDN |
| Gene Accession No. | D4AE48, Q3TPJ7, Q504T8 |
| Gene Alias | 3000003C15Rik; DKFZp547M072; midbrain nucleolar protein; Midn; Midnolin |
| Gene Symbols | MIDN |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.