Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MIER1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA540896
Description
Peptide sequence: RRTGDEKGVE AIPEGSHIKD NEQALYELVK CNFDTEEALR RLRF Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.
The association of hMI-ER1 with Sp1 represents a novel mechanism for the negative regulation of Sp1 target promoters. Results demonstrate that alternate use of a facultative intron regulates the subcellular localization of hMI-ER1 proteins and this may have important implications for hMI-ER1 function in the regulation of transcription. Repressor activity is due to interaction and recruitment of a trichostatin A-sensitive histone deacetylase 1 (HDAC1).
Specifications
MIER1 | |
Polyclonal | |
Unconjugated | |
MIER1 | |
4933425I22Rik; 5830411K19Rik; early response 1; er1; hMI-ER1; Kiaa1610; mesoderm induction early response 1 homolog; mesoderm induction early response 1 homolog (Xenopus laevis; mesoderm induction early response 1, transcriptional regulator; mesoderm induction early response protein 1; MIER1; MI-ER1; MIER1 transcriptional regulator; RGD1562337; RP5-944N15.1 | |
Rabbit | |
Protein A | |
RUO | |
57708 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8N108 | |
MIER1 | |
Synthetic peptide directed towards the middle region of human MIER1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction