Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MIER1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565607
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565607 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565607 Supplier Invitrogen™ Supplier No. PA565607
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GPTGGNKKPLH ADMDTNGYET DNLTTDPKLA HMTARNENDF DEKSERPAKR RRVNSNGKES PGSSEFFQEA VSHGKFEELE NTDD Highest antigen sequence identity to the following orthologs - mouse 89%, rat 87%.

The association of hMI-ER1 with Sp1 represents a novel mechanism for the negative regulation of Sp1 target promoters. Results demonstrate that alternate use of a facultative intron regulates the subcellular localization of hMI-ER1 proteins and this may have important implications for hMI-ER1 function in the regulation of transcription. Repressor activity is due to interaction and recruitment of a trichostatin A-sensitive histone deacetylase 1 (HDAC1).
TRUSTED_SUSTAINABILITY

Specifications

Antigen MIER1
Applications ChIP Assay, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MIER1
Gene Accession No. Q8N108
Gene Alias 4933425I22Rik; 5830411K19Rik; early response 1; er1; hMI-ER1; Kiaa1610; mesoderm induction early response 1 homolog; mesoderm induction early response 1 homolog (Xenopus laevis; mesoderm induction early response 1, transcriptional regulator; mesoderm induction early response protein 1; MIER1; MI-ER1; MIER1 transcriptional regulator; RGD1562337; RP5-944N15.1
Gene Symbols MIER1
Host Species Rabbit
Immunogen Recombinant Human MIER1. Recombinant protein control fragment (Product #RP-106272).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57708
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.