Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MIER2 Antibody (OTI3G7), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272683B
Description
MIER2 Monoclonal specifically detects MIER2 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
MIER2 | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
MIER2 | |
Recombinant protein fragment corresponding to amino acids 1-296 of human MIER2 (NP_060020) produced in E.coli. | |
0.1 mL | |
54531.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI3G7 | |
Western Blot, Immunohistochemistry | |
KIAA1193mesoderm induction early response protein 2, mesoderm induction early response 1, family member 2, Mi-er2 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction