Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MIER2 Monoclonal Antibody (OTI3A1)
GREENER_CHOICE

Catalog No. PIMA525903
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA525903 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA525903 Supplier Invitrogen™ Supplier No. MA525903
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MIER2 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)

Transcriptional repressor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MIER2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI3A1
Concentration 0.72 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Mier2
Gene Accession No. Q8N344
Gene Alias 2700087H15Rik; KIAA1193; mesoderm induction early response 1, family member 2; mesoderm induction early response protein 2; MIER family member 2; MIER2; Mi-er2; mKIAA1193; RGD1307278
Gene Symbols Mier2
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-296 of MIER2 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54531
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.