Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MIER2 Monoclonal Antibody (OTI3A1), TrueMAB™, OriGene
SDP

Catalog No. 502540345
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0345 30 μL
50-167-5103 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0345 Supplier OriGene Supplier No. TA504403S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MIER2 Monoclonal antibody specifically detects MIER2 in Human, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen MIER2
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI3A1
Concentration 0.72 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene Nln
Gene Accession No. P42676, Q9BYT8
Gene Alias 2700087H15Rik; KIAA1193; mesoderm induction early response 1, family member 2; mesoderm induction early response protein 2; MIER family member 2; MIER2; Mi-er2; mKIAA1193; RGD1307278
Host Species Mouse
Immunogen Full length human recombinant protein of human NLN produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117041, 57486
Target Species Human, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.