Learn More
Invitrogen™ MIER2 Monoclonal Antibody (OTI3G7)

Description
MIER2 Monoclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | MIER2 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI3G7 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | Mier2 |
| Gene Accession No. | Q8N344 |
| Gene Alias | 2700087H15Rik; KIAA1193; mesoderm induction early response 1, family member 2; mesoderm induction early response protein 2; MIER family member 2; MIER2; Mi-er2; mKIAA1193; RGD1307278 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.