Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MIER3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566161
Description
Immunogen sequence: HETSDFFPRPL RSNTACDGDK ESEVEDVETD SGNSPEDLRK EIMIGLQYQA EIPPYLGEYD GNEK Highest antigen sequence identity to the following orthologs - mouse 86%, rat 94%.
Transcriptional repressor.
Specifications
MIER3 | |
Polyclonal | |
Unconjugated | |
MIER3 | |
5730509D14; AU015422; D130064H19Rik; mesoderm induction early response 1, family member 3; mesoderm induction early response protein 3; MIER family member 3; Mier3; Mi-er3; RGD1311241 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
166968 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q7Z3K6 | |
MIER3 | |
Recombinant Human MIER3. Recombinant protein control fragment (Product #RP-106846). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction