Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MIF Antibody (4E4), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP104319AF594
Description
MIF Monoclonal specifically detects MIF in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
MIF | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
MIF | |
Recombinant human MIF (1-115 aa) purified from E. coli | |
0.1 mL | |
Primary | |
Human | |
IgG1 κ |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
40000 | |
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
EC 5.3.2.1, EC 5.3.3.12, GIFmacrophage migration inhibitory factor, GLIF, Glycosylation-inhibiting factor, L-dopachrome isomerase, L-dopachrome tautomerase, macrophage migration inhibitory factor (glycosylation-inhibiting factor), MMIF, Phenylpyruvate tautomerase | |
Mouse | |
Protein G purified | |
Biologically Active Proteins, Cytokine Research, Immunology, Innate Immunity | |
4282.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction