Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MIF Antibody (7A9), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | MIF |
---|---|
Clone | 7A9 |
Dilution | Western Blot 1:500-1:5000, ELISA |
Applications | Western Blot, ELISA |
Classification | Monoclonal |
Description
MIF Monoclonal antibody specifically detects MIF in Human samples. It is validated for Western Blot, ELISASpecifications
MIF | |
Western Blot 1:500-1:5000, ELISA | |
Monoclonal | |
Purified | |
RUO | |
Human | |
EC 5.3.2.1, EC 5.3.3.12, GIFmacrophage migration inhibitory factor, GLIF, Glycosylation-inhibiting factor, L-dopachrome isomerase, L-dopachrome tautomerase, macrophage migration inhibitory factor (glycosylation-inhibiting factor), MMIF, Phenylpyruvate tautomerase | |
A synthesized peptide derived from Human MIF [UniProt P14174] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
7A9 | |
Western Blot, ELISA | |
Unconjugated | |
Rabbit | |
Biologically Active Proteins, Cytokine Research, Immunology, Innate Immunity | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
4282 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title