Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human MIF Alexa Fluor 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC2891R100UG
This item is not returnable.
View return policy
Description
MIF Monoclonal specifically detects MIF in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
MIF | |
Monoclonal | |
Alexa Fluor 647 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
EC 5.3.2.1, EC 5.3.3.12, GIFmacrophage migration inhibitory factor, GLIF, Glycosylation-inhibiting factor, L-dopachrome isomerase, L-dopachrome tautomerase, macrophage migration inhibitory factor (glycosylation-inhibiting factor), MMIF, Phenylpyruvate tautomerase | |
E. coli-derived recombinant human MIF Met1-Phe114 Accession # P14174 | |
RUO | |
4282 | |
Human | |
IgG1 |
Flow Cytometry | |
932606 | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
P14174 | |
Mouse | |
100 μg | |
Primary | |
Detects human MIF in direct ELISAs. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction