Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MIF4GD Monoclonal Antibody (OTI2H9), TrueMAB™, OriGene
SDP

Catalog No. 502540253
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0253 30 μL
50-167-5216 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0253 Supplier OriGene Supplier No. TA504558S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MIF4GD Monoclonal antibody specifically detects MIF4GD in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot

Specifications

Antigen MIF4GD
Applications Flow Cytometry, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone OTI2H9
Concentration 0.68 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene NANP
Gene Accession No. Q8TBE9
Gene Alias 1110014L05Rik; 2310075G12Rik; AD023; hSLIP1; hypothetical protein LOC613305; MIF4G domain containing; MIF4G domain-containing protein; Mif4gd; MIFD; RGD1309685; SLBP (stem loop binding protein)-interacting protein 1; SLBP-interacting protein 1; SLC25A19; SLIP1; solute carrier family 25 (mitochondrial thiamine pyrophosphate carrier), member 19
Host Species Mouse
Immunogen Full length human recombinant protein of human NANP produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140838
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.