Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MIF4GD Monoclonal Antibody (OTI4C11)
GREENER_CHOICE

Catalog No. PIMA525913
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA525913 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA525913 Supplier Invitrogen™ Supplier No. MA525913
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MIF4GD Monoclonal Antibody for Western Blot, ICC/IF, Flow

MIF4GD is a protein which contains an MIF4G domain.This gene encodes a protein which contains an MIF4G domain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MIF4GD
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI4C11
Concentration 0.78 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MIF4GD
Gene Accession No. A9UHW6
Gene Alias 1110014L05Rik; 2310075G12Rik; AD023; hSLIP1; hypothetical protein LOC613305; MIF4G domain containing; MIF4G domain-containing protein; Mif4gd; MIFD; RGD1309685; SLBP (stem loop binding protein)-interacting protein 1; SLBP-interacting protein 1; SLC25A19; SLIP1; solute carrier family 25 (mitochondrial thiamine pyrophosphate carrier), member 19
Gene Symbols MIF4GD
Host Species Mouse
Immunogen Full length human recombinant protein of MIF4GD produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57409
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.