Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MIF4GD Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA562349
Description
Immunogen sequence: GEPSREEYKI QSFDAETQQL LKTALKDPGA VDLEKVANVI VDHSLQDCVF SKEAGRMCYA IIQAESKQAG QSVFRRGL Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 96%.
MIF4GD is a protein which contains an MIF4G domain.This gene encodes a protein which contains an MIF4G domain.
Specifications
MIF4GD | |
Polyclonal | |
Unconjugated | |
MIF4GD | |
1110014L05Rik; 2310075G12Rik; AD023; hSLIP1; hypothetical protein LOC613305; MIF4G domain containing; MIF4G domain-containing protein; Mif4gd; MIFD; RGD1309685; SLBP (stem loop binding protein)-interacting protein 1; SLBP-interacting protein 1; SLC25A19; SLIP1; solute carrier family 25 (mitochondrial thiamine pyrophosphate carrier), member 19 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57409 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
A9UHW6 | |
MIF4GD | |
Recombinant protein corresponding to Human MIF4GD. Recombinant protein control fragment (Product #RP-101252). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction