Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mifepristone, 98%, Thermo Scientific Chemicals

Catalog No. ac459982500
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84371-65-3
Molecular Formula C29H35NO2
Molecular Weight (g/mol) 429.59
InChI Key VKHAHZOOUSRJNA-GCNJZUOMSA-N
Synonym mifepristone, mifeprex, mifegyne, mifepriston, corlux, mifepristona, mifepristonum, korlym, mifepristonum latin, mifepristona spanish
PubChem CID 55245
ChEBI CHEBI:50692
IUPAC Name (8S,11R,13S,14S,17S)-11-[4-(dimethylamino)phenyl]-17-hydroxy-13-methyl-17-prop-1-ynyl-1,2,6,7,8,11,12,14,15,16-decahydrocyclopenta[a]phenanthren-3-one
SMILES CC#CC1(CCC2C1(CC(C3=C4CCC(=O)C=C4CCC23)C5=CC=C(C=C5)N(C)C)C)O

Specifications

Melting Point 192.0°C to 198.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 250 mg
Formula Weight 429.59
Percent Purity 98%
Chemical Name or Material Mifepristone
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May damage fertility or the unborn child.
Precautionary Statement GHS P Statement
Obtain special instructions before use.
Use personal protective equipment as required.
IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.