Learn More
Invitrogen™ Mig-6 Polyclonal Antibody

Description
The antibody detects endogenous levels of total Mig-6 protein.
Specifications
Specifications
| Antigen | Mig-6 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 2.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | Errfi1 |
| Gene Accession No. | Q9UJM3 |
| Gene Alias | 1300002F13Rik; 33; AI788755; ERBB receptor feedback inhibitor 1; Errfi1; gene 33; Gene 33 polypeptide; GENE-33; MIG6; MIG-6; Mitogen-inducible gene 6 protein; mitogen-inducible gene 6 protein homolog; RALT; Receptor-associated late transducer; RGD1307599; Xxx |
| Gene Symbols | Errfi1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.