Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mig-6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588982
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588982 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588982 Supplier Invitrogen™ Supplier No. PA588982
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SYVSDQNGGV PDPNPPPPQT HRRLRRSHSG PAGSFNKPAI RISNCCIHRA SPNSDEDKPE VPPRVPIPPR PVKPDYRRWS AEVTSSTYSD EDRPPKVPPR EPLSPSNSRT PSPKSLPSYL NGVMPPTQSF APDPKYVSSK ALQRQNSEGS ASKVPCILPI IENGKKVSST HYYLLPERPP YLDKYEKFFR EAEETNGGAQ IQPLPADCGI SSATEKPDSK TKMDLGGHVK RKHLSYVVSP; Positive Samples: Rat lung; Cellular Location: Cell membrane, Cytoplasm, Cytoplasmic side, Nucleus, Peripheral membrane protein.

ERRFI1 is a cytoplasmic protein whose expression is upregulated with cell growth (Wick et al., 1995). It shares significant homology with the protein product of rat gene-33, which is induced during cell stress and mediates cell signaling (Makkinje et al., 2000).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mig-6
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.43 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Errfi1
Gene Accession No. P05432
Gene Alias 1300002F13Rik; 33; AI788755; ERBB receptor feedback inhibitor 1; Errfi1; gene 33; Gene 33 polypeptide; GENE-33; MIG6; MIG-6; Mitogen-inducible gene 6 protein; mitogen-inducible gene 6 protein homolog; RALT; Receptor-associated late transducer; RGD1307599; Xxx
Gene Symbols Errfi1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 223-462 of human ERRFI1 (NP_0618211).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 313729
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.