Learn More
MIG 7 Polyclonal Antibody, Abbomax
Rabbit Polyclonal Antibody
Supplier: Abbomax Inc 602920

Description
Positive control: MCF-7 cells Cellular location: Membrane, Cytoplasmic.
MIG 7 Polyclonal antibody specifically detects MIG 7 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
MIG 7 | |
Polyclonal | |
Unconjugated | |
TOX4 | |
5730589K01Rik; A630040M18; AA410149; C14orf92; Epidermal Langerhans cell protein LCP1; KIAA0737; LCP1; MIG7; migration-inducing protein 7; RGD708449; TOX high mobility group box family member 4; TOX4; Unknown (protein for MGC:139878) | |
A synthetic peptide MAASRCSGLS corresponding to the N-terminus of human MIG-7 protein | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with proprietary stabilizer and 0.01% sodium azide | |
O94842 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9878 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.