Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Migfilin Antibody (OTI3G1), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272691R
Description
Migfilin Monoclonal specifically detects Migfilin in Human samples. It is validated for Western Blot.Specifications
Migfilin | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
FBLIM1 | |
Recombinant protein fragment corresponding to amino acids 1-242 of human Migfilin (NP_060026) produced in E.coli. | |
0.1 mL | |
54751.0 | |
Store at 4°C in the dark. |
Western Blot | |
OTI3G1 | |
Western Blot | |
CAL, DKFZp434G171, FBLP-1FBLP1CSX-associated LIM, filamin binding LIM protein 1, filamin-binding LIM protein 1, filamin-binding LIM protein-1, MIG2-interacting protein, Migfilin, mitogen-inducible 2 interacting protein, Mitogen-inducible 2-interacting protein | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction