Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Migfilin Mouse anti-Human, Clone: FBLIM1/4600, Novus Biologicals™

Mouse Monoclonal Antibody
$240.00 - $492.50
Specifications
Antigen | Migfilin |
---|---|
Clone | FBLIM1/4600 |
Concentration | 0.2 mg/ml |
Dilution | Western Blot 1-2 ug/ml, Immunofluorescence 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array |
Applications | Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Peptide Array |
Description
Migfilin Monoclonal antibody specifically detects Migfilin in Human samples. It is validated for Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Protein ArraySpecifications
Migfilin | |
0.2 mg/ml | |
Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Peptide Array | |
Unconjugated | |
Mouse | |
Human | |
Q8WUP2 | |
54751 | |
IgG1 κ | |
Protein A or G purified |
FBLIM1/4600 | |
Western Blot 1-2 ug/ml, Immunofluorescence 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
Monoclonal | |
Purified | |
RUO | |
10 mM PBS with 0.05% BSA | |
CAL, DKFZp434G171, FBLP-1FBLP1CSX-associated LIM, filamin binding LIM protein 1, filamin-binding LIM protein 1, filamin-binding LIM protein-1, MIG2-interacting protein, Migfilin, mitogen-inducible 2 interacting protein, Mitogen-inducible 2-interacting protein | |
Recombinant full-length human Migfilin protein (Uniprot: Q8WUP2) | |
Primary | |
Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title