Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MIIP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560869
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560869 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560869 Supplier Invitrogen™ Supplier No. PA560869
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SDTLALPRHC LLGWDIFPPK SEKSSAPRNL DLWSSVSAEA QHQKLSGTSS PFHPASPMQM LPPTPTWSVP QVPRPHVPRQ KP Highest antigen sequence identity to the following orthologs: Mouse - 59%, Rat - 53%.

MIIP inhibits glioma cells invasion and down-regulates adhesion-and motility-associated genes such as NFKB2 and ICAM1. MIIP exhibits opposing effects to IGFBP2 on cell invasion.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MIIP
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MIIP
Gene Accession No. Q5JXC2
Gene Alias AA553001; AI853621; D4Wsu114e; IGFBP2-binding protein; IGFBP-2-Binding Protein, IIp45; Iip45; invasion inhibitory protein 45; invasion-inhibitory protein 45; migration and invasion inhibitory protein; migration and invasion-inhibitory protein; MIIP
Gene Symbols MIIP
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MIIP. Recombinant protein control fragment (Product #RP-94086).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60672
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.