Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mili / PiwiL2 Monoclonal Antibody (17.8), Active Motif™
SDP

Catalog No. 501993180
Encompass
Change view
Click to view available options
Quantity:
10 μg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-3180 100 μg
50-199-3181 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-3180 Supplier Active Motif Supplier No. 61143
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The piwi gene of Drosophila belongs to a novel class of evolutionarily conserved genes (the piwi or Argonaute family). The Piwi family is required for germ and stem cell development in invertebrates, and two Piwi members, MIWI and MILI (Miwi like, Piwil2, Piwi-like protein 2), are essential for spermatogenesis in mouse. MIWI and MILI partner with Piwi-interacting RNAs (piRNAs) which are 26-31 nucleotides in length clearly distinct from the 21-23 nucleotides of microRNAs (miRNAs) or short interfering RNAs (siRNAs). A search for murine small RNAs that might program Piwi proteins for transposon suppression revealed developmentally regulated piRNA loci, some of which resemble transposon master control loci of Drosophila. MILI might be programmed by distinct piRNA populations at different stages of germ cell development, suggesting MILI plays an important role in meiotic prophase progression.

Specifications

Antigen Mili / PiwiL2
Applications Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 17.8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 30% glycerol and 0.035% sodium azide; pH 7.5
Gene Piwil2
Gene Accession No. Q8CDG1
Gene Alias 80kDa PIWIL2 short isoform; Cancer/testis antigen 80; CT80; HILI; Mili; Miwi like; Miwi likw; piwi like homolog 2; piwi like RNA-mediated gene silencing 2; Piwil1l; Piwil2; PIWI-L2; piwil2-like protein; piwi-like 2; piwi-like homolog 2; Piwi-like protein 2; piwi-like RNA-mediated gene silencing 2; testicular tissue protein Li 141
Gene Symbols Piwil2
Host Species Mouse
Immunogen This Mili / PiwiL2 antibody was raised against full-length recombinant mouse MILI / PIWIL2 protein.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57746
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.