Description
- Milky quartz mineral sample, 1 to 1.25 inches in size
- Naturally occurring white, opaque quartz
- Common form of silicon dioxide (SiO2)
- Ideal for classroom use, geology labs, or rock collections
- Great for identifying crystal structure and mineral properties
- Suitable for Grades 3–12 earth science curriculum
- Packaged for individual or group educational use
Specifications
Specifications
| Product Type | Mineral |