Learn More
Eisco™ Milky Quartz Mineral Specimen
SCI_ED
Catalog No. S26608 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1 Ea.
10/Pk.
12/Pk.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S26608 10/Pk.
S24710 1 Ea.
S24711 12/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S26608 Supplier Eisco™ Supplier No. ESNG0025PK10
Add to Cart
Add to Cart
Milky Quartz Mineral Specimen has a tightly packed framework and hardness of 7, complete lack of cleavage planes, and distinct euhedral crystal habit.
  • For use in classroom identification labs and exercises
  • For use in lessons on mineral hardness, luster, and fracture
  • Correlates well to Next Generation Science Standards Evidence Statements
  • Used in identification exercises, asking students to distinguish between it and other visually similar minerals such as feldspar or calcite
Specifications
Product Type Mineral
Quantity 10/Pk.
Specimen Size 2 to 3 cm
Weight (Metric) 0.2 kg
Specimen Type Milky Quartz