Learn More
StatLab™ Millennia™ Command Adhesion Microscope Slides
Click to view available options
Color:
Aqua
Blue
Green
Lilac
Pink
Red
Tan
White
Yellow
Description
- Simplify workflow and improve performance with Millennia Command Slides - a true all-purpose slide:
- A hybrid slide featuring a hydrophilic surface suitable for IHC, H&E and special staining
- Optimized coating for strong adhesion to reduce tissue loss
- Consistent staining with minimal background staining during all types of staining applications
- Compatible with thermal transfer printers
- Slide features 90-degree corners
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Sterility | Non-sterile |
| Edges | 90°, Ground |
| Frosted | 1 End, 1 Side |
| Precleaned | Yes |
| Type | Adhesion Slide |
| Material | High Quality Glass |
| Color | Pink |