Learn More
StatLab™ Millennia™ Command Adhesion Microscope Slides
Click to view available options
Color:
Aqua
Blue
Green
Lilac
Pink
Red
Tan
White
Yellow
Description
- Simplify workflow and improve performance with Millennia Command Slides - a true all-purpose slide:
- A hybrid slide featuring a hydrophilic surface suitable for IHC, H&E and special staining
- Optimized coating for strong adhesion to reduce tissue loss
- Consistent staining with minimal background staining during all types of staining applications
- Compatible with thermal transfer printers
- Slide features 90-degree corners
Specifications
Specifications
Precleaned | Yes |
Material | High Quality Glass |
Sterility | Non-sterile |
Edges | 90° |
Frosted | 1 End, 1 Side |
Type | Adhesion Slide |
Color | Tan |