Learn More
Honeywell™ Miller™ Adjustable-Fit Harnesses
Catalog No. 1798722A
Change view
Click to view available options
Friction buckle; X-Large
Tongue buckle; Large
Tongue buckle; Small
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
17-987-22A Tongue buckle; Small
17-987-22C Tongue buckle; Large
19-034-823D Friction buckle; X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 17-987-22A Supplier Honeywell™ Supplier No. 84284/SYK/DEL
Lightweight harnesses designed to arrest the most severe free falls. Constructed of high-visibility 1 3/4 in. (4.4cm) nylon webbing. Leg, waist, and shoulder straps adjust easily and fit comfortably. With friction waist buckle and shoulder buckle for custom fit. Sub-pelvic strap provides additional wearing comfort.
17-987-22 series has tongue-buckle leg straps for extra security; 19-034-823 has friction buckle.
TRUSTED_SUSTAINABILITY
Specifications
Size Small
Length (Metric) 4.4 cm
Buckle Style Tongue buckle
Length (English) 1.75 in.