Learn More
Honeywell™ Miller AirCore™ Oil and Gas Harness with Attached Bos'n Chair
Description
One harness for every job!
The Miller AirCore™ Oil and Gas Harness is built strong for day-to-day wear and tear and to last in a variety of harsh environments. This new line of AirCore harnesses keeps workers safe whether they are positioning pipe, or doing routine rig set up, required maintenance or tear down.
Certifications
Meets all applicable OSHA, ANSI and CSA standards
Specifications
Specifications
| Includes | Attached Bos'n Chair |
| D-Ring Location | Front, Extended back D-ring |
| Certifications/Compliance | Meets all applicable OSHA, ANSI and CSA standards |
| Buckle Style (Shoulder) | Cam buckles |
| Buckle Style (Leg) | Tongue buckle |
| Buckle Style (Chest) | Mating buckle |