Learn More
Honeywell™ Miller AirCore™ Oil and Gas Harness with Attached Seat Sling
Catalog No. 19066236 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Weight (English):
7.7 lb.
Catalog No. Weight (English)
19-066-236 7.7 lb.
1 options
Catalog No. 19-066-236 Supplier Honeywell™ Supplier No. ACOGTBSSUG
  • $2,005.00
    $1,140.65 / Each of 1
    Save $864.35
    43% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
The Miller AirCore™ Harness with Attached Sling Seat is designed for workers on drilling or service rigs. This harness features tongue buckle legs, mating buckle chest strap, front D-ring, extended back D-ring, lumbar pad, and removable belt with lumbar D-ring.
One harness for every job!
The Miller AirCore™ Oil and Gas Harness is built strong for day-to-day wear and tear and to last in a variety of harsh environments. This new line of AirCore harnesses keeps workers safe whether they are positioning pipe, or doing routine rig set up, required maintenance or tear down.
Certifications
Meets all applicable OSHA, ANSI and CSA standards
Specifications
Weight (English) 7.7 lb.
Includes Attached Seat Sling
Certifications/Compliance Meets all applicable OSHA, ANSI and CSA standards
Buckle Style (Shoulder) Cam buckle
D-Ring Location Front, Extended back D-ring
Buckle Style (Chest) Mating buckle
Buckle Style (Leg) Tongue buckle