Learn More
Honeywell™ Miller™ DuraFlex™ 650 Series Full-Body Stretchable Harnesses
Description
With comfort unsurpassed, DuraFlex Harnesses offer wider end-user acceptance. And, a comfortable harness is worn properly, enhancing compliance and safety on the job site.
- Maximum Comfort: harnesses conform to the shape of the worker, stretching with every movement, so there is no bunching, binding or kinking
- Increased Productivity: because DuraFlex Harnesses are stretchable, the worker has greater mobility with less fatigue
- Specially-formulated elastomer webbing stretches to provide greater comfort, improved safety and increased productivity
- Soft and flexible D-ring back pad further enhances comfort
- Self-contained label pack encapsulates labels, minimizing damage and loss
- Integrated and oversized pull-free lanyard rings
- Hazards: confined Space, fall from height
- Rated to 400 lb. capacity
Duraflex Harnesses
- 17-987-605 (Model E650-4/S/MGN): Friction buckle shoulder straps; tongue buckle legs straps; Small/Medium
- 19-046-150 (Model E650D-4/UBL): Friction buckle shoulder straps; tongue buckle legs straps; Blue; Universal
Duraflex Ultra Harnesses
- 19-274-710 (Model E650QC/UGN): Quick connect buckle chest strap and legs straps; Comfort D-pad; Universal
- 18-825-104 (Model E650QC-4/UGN): Quick connect buckle chest strap, tongue buckle legs straps; Comfort D-pad; Universal
- 18-825-105 (Model E650QC-4/S/MGN): Quick connect buckle chest strap, tongue buckle legs straps; Comfort D-pad; Small/Medium
Applications:
Construction, Manufacturing, Wind Energy
Compliance
Meets all applicable OSHA, ANSI and CSA standards
Warranty and Services
Limited lifetime, in the unlikely event that you should discover defects in either workmanship or materials. Does not apply to normal wear and tear or abusive treatment of the product.
Specifications
Specifications
| Webbing Material | Elastomer |
| Includes | Friction buckle shoulder straps; tongue buckle legs straps |
| Size | Small/Medium |
| Product Type | DuraFlex 650 Series Full-Body Stretchable Harness |