Learn More
Honeywell™ Miller™ DuraFlex™ 650 Series Full-Body Stretchable Harnesses
Catalog No. 18825105
Change view
Click to view available options
Includes:
Friction buckle shoulder straps; tongue buckle legs straps
Friction buckle shoulder straps; tongue buckle legs straps (blue)
Quick connect buckle chest strap and legs straps; Comfort D-pad
Quick connect buckle chest strap, tongue buckle legs straps; Comfort D-pad
Size:
Small/Medium
Universal
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes Size
18-825-105 Quick connect buckle chest strap, tongue buckle legs straps; Comfort D-pad Small/Medium
19-046-150 Friction buckle shoulder straps; tongue buckle legs straps (blue) Universal
17-987-605 Friction buckle shoulder straps; tongue buckle legs straps Small/Medium
19-274-710 Quick connect buckle chest strap and legs straps; Comfort D-pad Universal
18-825-104 Quick connect buckle chest strap, tongue buckle legs straps; Comfort D-pad Universal
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-825-105 Supplier Honeywell™ Supplier No. E650QC4/S/MGN
Feel the difference with specially-formulated elastomer (stretchable) webbing
With comfort unsurpassed, DuraFlex Harnesses offer wider end-user acceptance. And, a comfortable harness is worn properly, enhancing compliance and safety on the job site.
  • Maximum Comfort: harnesses conform to the shape of the worker, stretching with every movement, so there is no bunching, binding or kinking
  • Increased Productivity: because DuraFlex Harnesses are stretchable, the worker has greater mobility with less fatigue
  • Specially-formulated elastomer webbing stretches to provide greater comfort, improved safety and increased productivity
  • Soft and flexible D-ring back pad further enhances comfort
  • Self-contained label pack encapsulates labels, minimizing damage and loss
  • Integrated and oversized pull-free lanyard rings
  • Hazards: confined Space, fall from height
  • Rated to 400 lb. capacity
Duraflex Harnesses
  • 17-987-605 (Model E650-4/S/MGN): Friction buckle shoulder straps; tongue buckle legs straps; Small/Medium
  • 19-046-150 (Model E650D-4/UBL): Friction buckle shoulder straps; tongue buckle legs straps; Blue; Universal
Duraflex Ultra Harnesses
  • 19-274-710 (Model E650QC/UGN): Quick connect buckle chest strap and legs straps; Comfort D-pad; Universal
  • 18-825-104 (Model E650QC-4/UGN): Quick connect buckle chest strap, tongue buckle legs straps; Comfort D-pad; Universal
  • 18-825-105 (Model E650QC-4/S/MGN): Quick connect buckle chest strap, tongue buckle legs straps; Comfort D-pad; Small/Medium
Applications:
Construction, Manufacturing, Wind Energy
Compliance
Meets all applicable OSHA, ANSI and CSA standards
Warranty and Services
Limited lifetime, in the unlikely event that you should discover defects in either workmanship or materials. Does not apply to normal wear and tear or abusive treatment of the product.
TRUSTED_SUSTAINABILITY
Specifications
Webbing Material Elastomer
Product Type DuraFlex 650 Series Full-Body Stretchable Harness
Includes Quick connect buckle chest strap, tongue buckle legs straps; Comfort D-pad
Size Small/Medium