Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Miller™ DuraHoist™ Accessories

Catalog No. 19175447 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Product Type:
Floor Mount Sleeve
One piece adjustable mast
Material:
Aluminum
Zinc-plated Mild Steel
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Material
19-175-447 Floor Mount Sleeve Zinc-plated Mild Steel
19-175-445 One piece adjustable mast Aluminum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-175-447 Supplier Honeywell™ Supplier No. DH7ZP/
  • $2,207.00
    $1,617.65 / Each of 1

    Save $589.35

    27% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For use with DuraHoist Confined Space Systems

One-Piece Adjustable Mast

  • Mast offset adjusts from 12 to 29 in. (30 to 74cm)

Three-Piece Portable Base

  • Lightweight aluminum components; adjustable aluminum screws and a built-in level indicator
  • No tools required for assembly

Floor Mount Sleeve

  • Mounts to horizontal concrete or steel structures

Wall Mount Sleeve

  • Mounts to vertical concrete or steel structures

Flush Floor Mount Sleeve

  • Eliminates potential trip hazard
  • Installs into 4 in. dia. (10cm) core hole

Vehicle Hitch Mount Sleeve

  • Installs into 2 in. dia. (5cm) receiver on an attendant vehicle
  • Provides portable anchor point

Specifications

Material Zinc-plated Mild Steel
Product Type Floor Mount Sleeve
Load Bearing Capacity (Metric) 204 kg
Certifications/Compliance OSHA, ANSI, CSA
Load Bearing Capacity (English) 450 lb.
For Use With (Equipment) DuraHoist confined space system
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.