Learn More
Honeywell Safety Products™ Miller™ HP™ Fall Arrest and Retrieval Harnesses
Description
Superior water and oil repellency and protection against dirt, grime, spills and stains.
- With lanyard ring, sub-pelvic strap, back strap and back or side D-rings
- Bright green for high visibility and ease of inspection
- Friction buckles on the shoulder straps
(This style is also available with front D-ring, side D-rings and mating buckles on the shoulder straps. See below.) Models 850T and 850T-7 have mating buckles on both the leg and chest straps. Models 850T-4 and 850T-58 have tongue buckles on the leg straps and mating buckles on the chest strap.
Compliance
Meet all applicable OSHA, ANSI, and CSA requirements.
Specifications
Specifications
| Material | PTFE-coated Nylon Webbing |
| Size | Small/Medium |
| Buckle Style | Qwik-Fit |
| D-Ring Location | Back |
| Certifications/Compliance | Meets all applicable OSHA, ANSI, and CSA requirements. |