Learn More
Honeywell™ Miller™ Ms. Miller™ Duraflex Full-Body Harness
Catalog No. 19034405 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Color:
Green
Catalog No. Color
19-034-405 Green
1 options
Catalog No. 19-034-405 Supplier Honeywell™ Supplier No. E570/URN
  • $592.00
    $336.65 / Each of 1
    Save $255.35
    43% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
The 570/E570 Ms. Miller Harness is the only full-body harness on the market specifically designed for the female worker. The special design keeps shoulder straps at the side and away from the chest, offers better hip support and increases comfort.
  • Specially designed harnesses offer women greater comfort and improved support
  • Features front positioning loop, back and leg pads, and pull-free lanyard rings
  • Highly visible, bright solid color and stripes also help wearer orient harness for easy donning
  • Lightweight 1.75 in. shoulder, chest, back, and leg straps available in yellow, non-stretch polyester webbing or green, stretchable Duralite™ material for daylong comfort
Specifications
Color Green
Size Small/Medium
Length (English) 32 to 44 in.