Description
- Constructed of Millerhyde, a tough open weave nylon that is double laminated with vinyl
- Lightweight, yet durable
- Easily cleaned
- Each bag is equipped with convenient snap loops to attach to a belt
- Available in a variety of closures
- Color: Yellow with black straps
Specifications
Specifications
| Product Type | Respirator Bag |
| Closure Type | Snap |
| Color | Yellow with black straps |
| Material | Open-weave Nylon, Double-laminated with Vinyl |
| Includes | Snap loops to attach to a belt, full face |