Learn More
Honeywell™ Miller™ Revolution™ Aramid Fiber/Nomex Harnesses wa
Description
- Feature tongue buckle ring and a front D-ring made of Kevlar™ Aramid fibers
- Available with or without removable belt
- Exceed worker expectations in comfort, fit, ease-of-use, style, durability, compliance, flexibility and convenience
Specifications
Specifications
| D-Ring Location | Front |
| Product Type | Fall Protection Harness without removable belt |
| Buckle Style (Leg) | Tongue buckle |
| Size | 2X-Large/3X-Large |
| Webbing Material | Nomex |