Learn More
Honeywell™ Miller™ Revolution™ Arc-Rated Harnesses
Catalog No. 19169922 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Product Type:
Arc-Rated Harness with Rescue Loops
Arc-Rated Harness without Rescue Loops
Size:
Small/Medium
Universal
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Size
19-169-922 Arc-Rated Harness with Rescue Loops Universal
17-987-606 Arc-Rated Harness without Rescue Loops Small/Medium
19-169-921 Arc-Rated Harness without Rescue Loops Universal
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-169-922 Supplier Honeywell™ Supplier No. RKNARRLQC/UBK
Add to Cart
Add to Cart
This item is not returnable. View return policy
Protect from falls caused by electric arc flash and arc blast exposure
  • Kevlar™/Nomex™ DualTech™ webbing has 10,000 lb. tensile strength; engineered with permanent fire-retardant properties
  • Kevlar outer offers durability and fire resistance while Nomex inner gives greater comfort
  • PivotLink™ design allows freedom of motion while cam buckles provide easy, two-finger adjustment
  • Feature quick-connect chest and leg buckles, cam buckles, a self-contained label pack, web finials, a Kevlar web back D-ring, and an ErgoArmor™ back shield
  • Leather insulators under all-metal hardware increase worker protection
  • Bright red binding on every pad for easy identification
Order Info
Universal size. Harnesses are rated to 400 lb. capacity.
Certifications
Meet ASTM™ F887-05 arc-flash standard.
Compliance
Meets OSHA 1910.66 and OSHA 1926.502 (d); ANSI™ A10.32 and ANSI Z359-2007; ASTM™ F887-13; CSA Z259.10 requirements.
Specifications
Material Dupont™ Kevlar™ aramid fiber, Nomex™ aramid fibers
Product Type Arc-Rated Harness with Rescue Loops
For Use With (Application) Designed to protect workers from falls caused by electric arc-flash and arc-blast exposure
Size Universal