Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Miller™ Revolution™ DualTech™ Harnesses

Catalog No. 19165302
Change view
Click to view available options
D-Ring Location:
Front
Front, Side
Includes:
Integrated accessory system
Pad, integrated accessory system
Removable belt and pad, integrated accessory system
Size:
2X-Large/3X-Large
Small/Medium
Universal
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. D-Ring Location Includes Size
19-165-302 Front, Side Removable belt and pad, integrated accessory system Small/Medium
19-165-283 Front Integrated accessory system Universal
19-165-292 Front, Side Removable belt and pad, integrated accessory system Universal
19-165-294 Front, Side Removable belt and pad, integrated accessory system 2X-Large/3X-Large
19-165-285 Front Integrated accessory system 2X-Large/3X-Large
19-165-289 Front, Side Pad, integrated accessory system Universal
19-165-290 Front, Side Pad, integrated accessory system Small/Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 19-165-302 Supplier Honeywell™ Supplier No. RDTFDTBBDP/S/MBK

Harnesses with DualTech webbing offering safety and functionality with features developed to meet key user needs

  • Available with tongue buckle legs and front and/or side D-rings
  • Available with or without removable belt
  • Exceed worker expectations in comfort, fit, ease-of-use, style, durability, compliance, flexibility and convenience
TRUSTED_SUSTAINABILITY

Specifications

Color Black
Includes Removable belt and pad, integrated accessory system
Certifications/Compliance OSHA, ANSI and CSA
Buckle Style (Leg) Tongue-Buckle Leg Straps
For Use With (Application) Construction, manufacturing and transportation fall protection
Webbing Material DualTech™, Polyester
Style Full-body harness
D-Ring Location Front, Side
Size Small/Medium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.