Learn More
Honeywell™ Miller™ Revolution™ Harness with Aramid Fiber-Nomex™ Webbing
Catalog No. 17987615 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
2X-Large; 3X-Large
Universal
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-987-615 Universal
17-987-616 2X-Large; 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-987-615 Supplier Honeywell™ Supplier No. RKNARFDQC/UBK
Add to Cart
Add to Cart
The Revolution Arc Rated Products are designed to protect workers from falls caused by electric arc-flash and arc-blast exposure when working at heights or in confined space applications near energized electrical sources.
Kevlar™ Aramid fibers/Nomex™ aramid fibers Miller DualTech Webbing. 10,000 lb. tensile strength webbing is engineered with permanent fire retardant (FR) properties. The webbing offers the durability and FR properties of Kevlar™ Aramid fibers on the outer side, with softer FR Nomex™ aramid fibers on the inside of the webbing for greater comfort. It assures outstanding arc protection and durability.
  • Arc rated, Quick-Connect buckle legs, front D-ring
  • Leather insulators under all metal hardware offer additional protection for the worker
  • The Kevlar™ Aramid fibers web loop on the back pad allows for a metal-less connection when used with a Kevlar™ Aramid fibers shock-absorbing lanyard with a choke-off loop
  • All Miller Revolution Arc-Rated Harnesses have bright red binding on every pad, making them easily identifiable as an arc-rated product
  • Hazards: Confined Space, Electrical and Arc, Fall from height, Welding Flash
  • Rated to 400 lb. capacity
  • Applications: Construction, Utilities
Compliance
Meets all applicable OSHA, ANSI and CSA standards
Specifications
Material Dupont™ Kevlar™ aramid fiber, Nomex™ aramid fibers
Size Universal
D-Ring Location Front
For Use With (Application) To protect worker from fall caused by electric arc-flash, arc-blast exposure when working at heights/energized electrical source