Learn More
Honeywell™ Miller™ Revolution™ Harness with Aramid Fiber-Nomex™ Webbing
Catalog No. 17987616 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
2X-Large; 3X-Large
Universal
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-987-616 2X-Large; 3X-Large
17-987-615 Universal
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-987-616 Supplier Honeywell™ Supplier No. RKNARFDQCBDP/3XLBK
  • $2,760.00
    $1,569.65 / Each of 1
    Save $1,190.35
    43% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
The Revolution Arc Rated Products are designed to protect workers from falls caused by electric arc-flash and arc-blast exposure when working at heights or in confined space applications near energized electrical sources.
Kevlar™ Aramid fibers/Nomex™ aramid fibers Miller DualTech Webbing. 10,000 lb. tensile strength webbing is engineered with permanent fire retardant (FR) properties. The webbing offers the durability and FR properties of Kevlar™ Aramid fibers on the outer side, with softer FR Nomex™ aramid fibers on the inside of the webbing for greater comfort. It assures outstanding arc protection and durability.
  • Arc rated, Quick-Connect buckle legs, front D-ring
  • Leather insulators under all metal hardware offer additional protection for the worker
  • The Kevlar™ Aramid fibers web loop on the back pad allows for a metal-less connection when used with a Kevlar™ Aramid fibers shock-absorbing lanyard with a choke-off loop
  • All Miller Revolution Arc-Rated Harnesses have bright red binding on every pad, making them easily identifiable as an arc-rated product
  • Hazards: Confined Space, Electrical and Arc, Fall from height, Welding Flash
  • Rated to 400 lb. capacity
  • Applications: Construction, Utilities
Compliance
Meets all applicable OSHA, ANSI and CSA standards
Specifications
Material Dupont™ Kevlar™ aramid fiber, Nomex™ aramid fibers
D-Ring Location Front
For Use With (Application) To protect worker from fall caused by electric arc-flash, arc-blast exposure when working at heights/energized electrical source
Size 2X-Large; 3X-Large